MilliporeSigma
  • Functional poly(ε-caprolactone)/chitosan dressings with nitric oxide-releasing property improve wound healing.
Functional poly(ε-caprolactone)/chitosan dressings with nitric oxide-releasing property improve wound healing.
Acta biomaterialia (2017-03-13)
Xin Zhou, He Wang, Jimin Zhang, Xuemei Li, Yifan Wu, Yongzhen Wei, Shenglu Ji, Deling Kong, Qiang Zhao
ABSTRACT
Wound healing dressings are increasingly needed clinically due to the large number of skin damage annually. Nitric oxide (NO) plays a key role in promoting wound healing, thus biomaterials with NO-releasing property receive increasing attention as ideal wound dressing. In present study, we prepared a novel functional wound dressing by combining electrospun poly(ε-caprolactone) (PCL) nonwoven mat with chitosan-based NO-releasing biomaterials (CS-NO). As-prepared PCL/CS-NO dressing released NO sustainably under the physiological conditions, which was controlled by the catalysis of β-galactosidase. In vivo wound healing characteristics were further evaluated on full-thickness cutaneous wounds in mice. Results showed that PCL/CS-NO wound dressings remarkably accelerated wound healing process through enhancing re-epithelialization and granulation formation and effectively improved the organization of regenerated tissues including epidermal-dermal junction, which could be ascribed to the pro-angiogenesis, immunomodulation, and enhanced collagen synthesis provided by the sustained release of NO. Therefore, PCL/CS-NO may be a promising candidate for wound dressings, especially for the chronic wound caused by the ischemia. Serious skin damage caused by trauma, surgery, burn or chronic disease has become one of the most serious clinical problems. Therefore, there is an increasing demand for ideal wound dressing that can improve wound healing. Due to the vital role of nitric oxide (NO), we developed a novel functional wound dressing by combining electrospun polycaprolactone (PCL) mat with NO-releasing biomaterial (CS-NO). The sustained release of NO from PCL/CS-NO demonstrated positive effects on wound healing, including pro-angiogenesis, immunomodulation, and enhanced collagen synthesis. Hence, wound healing process was remarkably accelerated and the organization of regenerated tissues was effectively improved as well. Taken together, PCL/CS-NO dressing may be a promising candidate for wound treatment, especially for the chronic wound caused by the ischemia.
Sigma-Aldrich
4-Pentynoic acid, 95%
Pricing and availability is not currently available.