MilliporeSigma
  • Chromatin recruitment of activated AMPK drives fasting response genes co-controlled by GR and PPARα.
Chromatin recruitment of activated AMPK drives fasting response genes co-controlled by GR and PPARα.
Nucleic acids research (2016-09-01)
Dariusz Ratman, Viacheslav Mylka, Nadia Bougarne, Michal Pawlak, Sandrine Caron, Nathalie Hennuyer, Réjane Paumelle, Lode De Cauwer, Jonathan Thommis, Mark H Rider, Claude Libert, Sam Lievens, Jan Tavernier, Bart Staels, Karolien De Bosscher
ABSTRACT
Adaptation to fasting involves both Glucocorticoid Receptor (GRα) and Peroxisome Proliferator-Activated Receptor α (PPARα) activation. Given both receptors can physically interact we investigated the possibility of a genome-wide cross-talk between activated GR and PPARα, using ChIP- and RNA-seq in primary hepatocytes. Our data reveal extensive chromatin co-localization of both factors with cooperative induction of genes controlling lipid/glucose metabolism. Key GR/PPAR co-controlled genes switched from transcriptional antagonism to cooperativity when moving from short to prolonged hepatocyte fasting, a phenomenon coinciding with gene promoter recruitment of phosphorylated AMP-activated protein kinase (AMPK) and blocked by its pharmacological inhibition. In vitro interaction studies support trimeric complex formation between GR, PPARα and phospho-AMPK. Long-term fasting in mice showed enhanced phosphorylation of liver AMPK and GRα Ser211. Phospho-AMPK chromatin recruitment at liver target genes, observed upon prolonged fasting in mice, is dampened by refeeding. Taken together, our results identify phospho-AMPK as a molecular switch able to cooperate with nuclear receptors at the chromatin level and reveal a novel adaptation mechanism to prolonged fasting.
Product Description
SAFC
Williams′ Medium E, With sodium bicarbonate, without L-glutamine and phenol red, liquid, sterile-filtered, suitable for cell culture