MilliporeSigma
  • Thymoquinone binds and activates human salivary aldehyde dehydrogenase: Potential therapy for the mitigation of aldehyde toxicity and maintenance of oral health.
Thymoquinone binds and activates human salivary aldehyde dehydrogenase: Potential therapy for the mitigation of aldehyde toxicity and maintenance of oral health.
International journal of biological macromolecules (2017-05-05)
Amaj Ahmed Laskar, Masood Alam Khan, Fizza Askari, Hina Younus
ABSTRACT
Human salivary aldehyde dehydrogenase (hsALDH) is a very important anti-oxidant enzyme present in the saliva. It is involved in the detoxification of toxic aldehydes and maintenance of oral health. Reduced level of hsALDH activity is a risk factor for oral cancer development. Thymoquinone (TQ) has many pharmacological activities and health benefits. This study aimed to examine the activation of hsALDH by TQ. The effect of TQ on the activity and kinetics of hsALDH was studied. The binding of TQ with the enzyme was examined by different biophysical methods and molecular docking analysis. TQ enhanced the dehydrogenase activity of crude and purified hsALDH by 3.2 and 2.9 fold, respectively. The K
Sigma-Aldrich
6-Methoxy-2-naphthaldehyde, 98%