MilliporeSigma
  • Identification of a HLA-A*0201-restricted immunogenic epitope from the universal tumor antigen DEPDC1.
Identification of a HLA-A*0201-restricted immunogenic epitope from the universal tumor antigen DEPDC1.
Oncoimmunology (2017-09-19)
Anna Tosi, Silvia Dalla Santa, Elisa Cappuzzello, Carolina Marotta, Dawid Walerich, Giannino Del Sal, Paola Zanovello, Roberta Sommaggio, Antonio Rosato
ABSTRACT
The identification of universal tumor-specific antigens shared between multiple patients and/or multiple tumors is of great importance to overcome the practical limitations of personalized cancer immunotherapy. Recent studies support the involvement of DEPDC1 in many aspects of cancer traits, such as cell proliferation, resistance to induction of apoptosis and cell invasion, suggesting that it may play key roles in the oncogenic process. In this study, we report that DEPDC1 expression is upregulated in most types of human tumors, and closely linked to a poorer prognosis; therefore, it might be regarded as a novel universal oncoantigen potentially suitable for targeting many different cancers. In this regard, we report the identification of a HLA-A*0201 allele-restricted immunogenic DEPDC1-derived epitope, which is able to induce cytotoxic T lymphocytes (CTL) exerting a strong and specific functional response
Sigma-Aldrich
MISSION® esiRNA, targeting human DEPDC1