MilliporeSigma
  • MicroRNA-128-3p regulates mitomycin C-induced DNA damage response in lung cancer cells through repressing
MicroRNA-128-3p regulates mitomycin C-induced DNA damage response in lung cancer cells through repressing
Oncotarget (2017-09-25)
Rui Zhang, Chang Liu, Yahan Niu, Ying Jing, Haiyang Zhang, Jin Wang, Jie Yang, Ke Zen, Junfeng Zhang, Chen-Yu Zhang, Donghai Li
ABSTRACT
The DNA damage response is critical for maintaining genome integrity and preventing damage to DNA due to endogenous and exogenous insults. Mitomycin C (MMC), a potent DNA cross-linker, is used as a chemotherapeutic agent because it causes DNA inter-strand cross-links (DNA ICLs) in cancer cells. While many microRNAs, which may serve as oncogenes or tumor suppressors, are grossly dysregulated in human cancers, little is known about their roles in MMC-treated lung cancer. Here, we report that miR-128-3p can attenuate repair of DNA ICLs by targeting
Sigma-Aldrich
Magnesium methyl carbonate solution, 2.0 M in DMF
Sigma-Aldrich
MISSION® esiRNA, targeting human SPTAN1