MilliporeSigma
A snake venom group IIA PLA
Scientific reports (2017-06-24)
Karina Cristina Giannotti, Elbio Leiguez, Ana Eduarda Zulim de Carvalho, Neide Galvão Nascimento, Márcio Hideki Matsubara, Consuelo Latorre Fortes-Dias, Vanessa Moreira, Catarina Teixeira
ABSTRACT
Crotoxin B (CB) is a catalytically active group IIA sPLA
Sigma-Aldrich
Bromoenol lactone, ≥98% (TLC)
Pricing and availability is not currently available.
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide, ≥97.0% (T)
Pricing and availability is not currently available.
Sigma-Aldrich
Valeryl Salicylate, ≥98% (HPLC)
Pricing and availability is not currently available.