MilliporeSigma
  • Fluorescent organic single crystals with elastic bending flexibility: 1,4-bis(thien-2-yl)-2,3,5,6-tetrafluorobenzene derivatives.
Fluorescent organic single crystals with elastic bending flexibility: 1,4-bis(thien-2-yl)-2,3,5,6-tetrafluorobenzene derivatives.
Scientific reports (2017-08-27)
Shotaro Hayashi, Atsushi Asano, Natsumi Kamiya, Yoshinobu Yokomori, Takuto Maeda, Toshio Koizumi
ABSTRACT
Organic single crystals with elastic bending flexibility are rare because they are generally brittle. We report here fluorescent organic single crystals based on thiophene-tetrafluorobenzene-thiophene derivatives, mainly 1,4-bis(thien-2-yl)-2,3,5,6-tetrafluorobenzene. Three derivatives were synthesized by Pd-catalyzed cross-coupling reactions (Stille or direct arylation pathways). The crystallization of the derivatives gave large (mm- or cm-scale) crystals. Two crystals of 1,4-bis(thien-2-yl)-2,3,5,6-tetrafluorobenzene, 1, and 1,4-bis(4-methylthien-2-yl)-2,3,5,6-tetrafluorobenzene, 3, bent under applied stress and quickly recovered its original shape upon relaxation. The other crystal of 1,4-bis(5-methylthien-2-yl)-2,3,5,6-tetrafluorobenzene, 2, showed brittle breakage under applied stress (normal behavior). Fibril lamella crystal structure based on criss-cross packed slip-stacked molecular wires and its structural integrity are important factors for the design and production of next generation crystal materials with elastic bending flexibility. Furthermore, mechanical bending-relaxation resulted in reversible change of the morphology and fluorescence (mechanofluorochromism). Such bendable crystals would lead to the next generation solid-state fluorescent and/or semiconducting materials.
Sigma-Aldrich
1-Adamantanecarboxylic acid, 99%