MilliporeSigma
  • The relative abundance of dimethylsulfoniopropionate (DMSP) among other zwitterions in branching coral at Heron Island, southern Great Barrier Reef.
The relative abundance of dimethylsulfoniopropionate (DMSP) among other zwitterions in branching coral at Heron Island, southern Great Barrier Reef.
Analytical and bioanalytical chemistry (2017-05-21)
Hilton B Swan, Elisabeth S M Deschaseaux, Graham B Jones, Bradley D Eyre
ABSTRACT
Dimethylsulfoniopropionate (DMSP) and eleven other target zwitterions were quantified in the branch tips of six Acropora species and Stylophora pistillata hard coral growing on the reef flat surrounding Heron Island in the southern Great Barrier Reef (GBR), Australia. Hydrophilic interaction liquid chromatography mass spectrometry (HILIC-MS) was used for sample analysis with isotope dilution MS applied to quantify DMSP. The concentration of DMSP was ten times greater in A. aspera than A. valida, with this difference being maintained throughout the spring, summer and winter seasons. In contrast, glycine betaine was present in significantly higher concentrations in these species during the summer than the winter. Exposure of branch tips of A. aspera to air and hypo-saline seawater for up to 1 h did not alter the concentrations of DMSP present in the coral when compared with control samples. DMSP was the most abundant target zwitterion in the six Acropora species examined, ranging from 44-78% of all target zwitterions in A. millepora and A. aspera, respectively. In contrast, DMSP only accounted for 7% in S. pistillata, with glycine betaine and stachydrine collectively accounting for 88% of all target zwitterions in this species. The abundance of DMSP in the six Acropora species examined points to Acropora coral being an important source for the biogeochemical cycling of sulfur throughout the GBR, since this reef-building branching coral dominates the coral cover of the GBR. Graphical Abstract HILIC-MS extracted ion chromatogram showing zwitterionic metabolites from the branching coral Acropora isopora.