MilliporeSigma
  • Leaf Extracts of Selected Gardening Trees Can Attenuate Quorum Sensing and Pathogenicity of
Leaf Extracts of Selected Gardening Trees Can Attenuate Quorum Sensing and Pathogenicity of
Indian journal of microbiology (2017-09-15)
Kaimin Niu, Min Kuk, Haein Jung, Kokgan Chan, Sooki Kim
ABSTRACT
An increasing concern on resistance to multiple-antibiotics has led to the discovery of novel agents and the establishment of new precaution strategy. Numerous plant sources have been widely studied to reduce virulence of pathogenic bacteria by interfering cell-to-cell based communication called quorum sensing (QS). Leaf extracts of 17 gardening trees were collected and investigated for their anti-QS effects using a sensor strain
Sigma-Aldrich
N-Heptanoyl-L-homoserine lactone, ≥96% (HPLC)
Sigma-Aldrich
Dihydro-3-amino-2-(3H)-furanone, AldrichCPR
Sigma-Aldrich
N-Hexanoyl-L-homoserine lactone, ≥96% (HPLC)