MilliporeSigma
  • How does live yeast differ from sodium bicarbonate to stabilize ruminal pH in high-yielding dairy cows?
How does live yeast differ from sodium bicarbonate to stabilize ruminal pH in high-yielding dairy cows?
Journal of dairy science (2008-09-04)
J P Marden, C Julien, V Monteils, E Auclair, R Moncoulon, C Bayourthe
ABSTRACT
The objectives of this study were to evaluate the capacity of 2 dietary feed additives, sodium bicarbonate and live yeast Saccharomyces cerevisiae (strain Sc 47), in optimizing ruminal pH in dairy cows and to determine their modes of action. Three early lactating Holstein cows, fitted with ruminal cannulas, were allocated in a 3 x 3 Latin square design. They were given a total mixed ration as control diet (CD) at a daily feeding rate of 28.0 kg of dry matter (DM)/cow supplemented with 150 g/d of sodium bicarbonate (SBD) or 5 g/d of live yeast (YD) during a 21-d experimental period (14 d of diet adaptation, 4 consecutive days of measurement and sampling and 3 d of transition). The pH and redox potential (E(h)) were measured from 1 h before feeding to 8 h after feeding at 1-h intervals, and samples of ruminal fluid were taken at 0, 2, 4, 6, and 8 h after feeding for the determination of volatile fatty acids and lactate concentrations. Total tract apparent digestibility of the diet was also determined. Ruminal pH fluctuated between 6.53 at feeding and 5.57 at 5 h postfeeding. Mean pH was greater with SBD (6.21) and YD (6.14) compared with CD (5.94), showing that both additives had a pH stabilization effect. The E(h) varied from -88 mV at 1 h before feeding to -165 mV at 1 h after feeding. Mean E(h) and Clark's Exponent (rH) were lower with YD (-149 mV and 7.31, respectively) than with SBD (-137 mV and 7.85, respectively) and CD (-115 mV and 8.05, respectively), indicating that the yeast strengthened the reducing power of the milieu. Total volatile fatty acids were greater in SBD (95.3 mM) and YD (99.4 mM) compared with CD (85.3 mM). Acetate concentration was greater in SBD (60.8 mM) and YD (59.1 mM) compared with CD (53.2 mM). Propionate concentration was greater in YD (25.8 mM) than in SBD (20.0 mM) and CD (18.0 mM). Butyrate remained constant between diets. Mean total lactate concentrations were 16.5, 12.2, and 5.4 mM for CD, SBD, and YD, respectively, with a 67% decrease with YD. Total tract organic matter digestibility was greater for YD (66.6%) compared with SBD (61.7%) and CD (62.2%). The neutral detergent fiber digestibility was greater with YD (41.6%) compared with SBD (34.3%) and CD (29.6%), whereas acid detergent fiber digestibility was greatest in YD (32.3%), intermediate in SBD (24.4%), and lowest in CD (18.1%). By inducing a lower ruminal E(h) and rH, live yeast prevented accumulation of lactate and allowed better fiber digestion, whereas sodium bicarbonate seemed to act only as an exogenous buffer.
Sigma-Aldrich
α-Amylase, heat-stable, solution, for use in Total Dietary Fiber Assay, TDF-100A