MilliporeSigma
  • Liquid-phase oxidation of alkylaromatics to aromatic ketones with molecular oxygen over a Mn-based metal-organic framework.
Liquid-phase oxidation of alkylaromatics to aromatic ketones with molecular oxygen over a Mn-based metal-organic framework.
Dalton transactions (Cambridge, England : 2003) (2017-06-18)
Yasutaka Kuwahara, Yukihiro Yoshimura, Hiromi Yamashita
ABSTRACT
Liquid-phase oxidation of alkylaromatics with molecular O
Sigma-Aldrich
2,5-Dihydroxyterephthalic acid, 98%