MilliporeSigma
  • An extended π-system and enhanced electronic delocalization on symmetric [Ru
An extended π-system and enhanced electronic delocalization on symmetric [Ru
Dalton transactions (Cambridge, England : 2003) (2017-06-13)
Bruna Possato, Victor M Deflon, Zeki Naal, André L B Formiga, Sofia Nikolaou
ABSTRACT
We report on the investigation of a new series of symmetric trinuclear ruthenium complexes combined with azanaphthalene ligands: [Ru
Sigma-Aldrich
Quinazoline, 99%