MilliporeSigma
  • Enhancing MRI of liver metastases with a zwitterionized biodegradable dendritic contrast agent.
Enhancing MRI of liver metastases with a zwitterionized biodegradable dendritic contrast agent.
Biomaterials science (2017-04-20)
Xiaoxuan Zhou, Mingzhou Ye, Yuxin Han, Jianbin Tang, Yue Qian, Hongjie Hu, Youqing Shen
ABSTRACT
Metastasis is the main reason for cancer-associated mortality, and accurate diagnostic imaging of metastases is critical for the clinical administration and tailoring personalized treatments for metastatic tumors. However, magnetic resonance imaging of metastases in the liver is impeded by its low sensitivity because the currently used contrast agents accumulate in hepatocytes and Kupffer cells instead of cancer cells. Herein, a 4
Sigma-Aldrich
2-tert-Butyl-4-methylphenol, 99%