MilliporeSigma
Thermoelectric performance of n-type Mg
Scientific reports (2017-06-24)
Rafael Santos, Mitchell Nancarrow, Shi Xue Dou, Sima Aminorroaya Yamini
ABSTRACT
Magnesium-based thermoelectric materials (Mg