MilliporeSigma
Structural studies of viperin, an antiviral radical SAM enzyme.
Proceedings of the National Academy of Sciences of the United States of America (2017-06-14)
Michael K Fenwick, Yue Li, Peter Cresswell, Yorgo Modis, Steven E Ealick
ABSTRACT
Viperin is an IFN-inducible radical
Sigma-Aldrich
2-Aminoimidazole, AldrichCPR
Sigma-Aldrich
Pyrimidine, ≥98.0%