MilliporeSigma
  • Amoeba-like self-oscillating polymeric fluids with autonomous sol-gel transition.
Amoeba-like self-oscillating polymeric fluids with autonomous sol-gel transition.
Nature communications (2017-07-14)
Michika Onoda, Takeshi Ueki, Ryota Tamate, Mitsuhiro Shibayama, Ryo Yoshida
ABSTRACT
In the field of polymer science, many kinds of polymeric material systems that show a sol-gel transition have been created. However, most systems are unidirectional stimuli-responsive systems that require physical signals such as a change in temperature. Here, we report on the design of a block copolymer solution that undergoes autonomous and periodic sol-gel transition under constant conditions without any on-off switching through external stimuli. The amplitude of this self-oscillation of the viscosity is about 2,000 mPa s. We also demonstrate an intermittent forward motion of a droplet of the polymer solution synchronized with the autonomous sol-gel transition. This polymer solution bears the potential to become the base for a type of slime-like soft robot that can transform its shape kaleidoscopically and move autonomously, which is associated with the living amoeba that moves forward by a repeated sol-gel transition.
Sigma-Aldrich
N-(3-Aminopropyl)methacrylamide hydrochloride, contains ≤1,000 ppm MEHQ as stabilizer, 98% (HPLC)