MilliporeSigma
  • Excited-State Deactivation Pathways and the Photocyclization of BN-Doped Polyaromatics.
Excited-State Deactivation Pathways and the Photocyclization of BN-Doped Polyaromatics.
The journal of physical chemistry. A (2017-06-20)
Joshua A Snyder, Peter Grüninger, Holger F Bettinger, Arthur E Bragg
ABSTRACT
Boron-nitrogen doping of polyaromatic hydrocarbons (PAH), such as borazine-core hexabenzocoronene, presents possibilities for tuning the properties of organic electronics and nanographene materials while preserving structural characteristics of pure hydrocarbons. Previous photochemical studies have demonstrated extension of a borazine-core PAH network (1,2:3,4:5,6-tris(o,o'-biphenylylene)borazine, 1) by photoinduced cyclodehydrogenation. We present steady-state and femtosecond-to-microsecond resolved spectroscopic studies of the photophysics of 1 and a related borazine-core PAH in order to characterize competing excited-state relaxation pathways that determine the efficacy of bond formation by photocyclization. Transient spectra evolve on time scales consistent with S
Sigma-Aldrich
Molecular sieves, 3 Å, pellets, ~1.6 mm
Pricing and availability is not currently available.