MilliporeSigma
  • Wafer-scale single-crystal perovskite patterned thin films based on geometrically-confined lateral crystal growth.
Wafer-scale single-crystal perovskite patterned thin films based on geometrically-confined lateral crystal growth.
Nature communications (2017-07-12)
Lynn Lee, Jangmi Baek, Kyung Sun Park, Yong-EunKoo Lee, Nabeen K Shrestha, Myung M Sung
ABSTRACT
We report a facile roll-printing method, geometrically confined lateral crystal growth, for the fabrication of large-scale, single-crystal CH
Sigma-Aldrich
2-Hydroxy-2-methylpropiophenone, 97%