MilliporeSigma
  • Wearable smart sensor systems integrated on soft contact lenses for wireless ocular diagnostics.
Wearable smart sensor systems integrated on soft contact lenses for wireless ocular diagnostics.
Nature communications (2017-04-28)
Joohee Kim, Minji Kim, Mi-Sun Lee, Kukjoo Kim, Sangyoon Ji, Yun-Tae Kim, Jihun Park, Kyungmin Na, Kwi-Hyun Bae, Hong Kyun Kim, Franklin Bien, Chang Young Lee, Jang-Ung Park
ABSTRACT
Wearable contact lenses which can monitor physiological parameters have attracted substantial interests due to the capability of direct detection of biomarkers contained in body fluids. However, previously reported contact lens sensors can only monitor a single analyte at a time. Furthermore, such ocular contact lenses generally obstruct the field of vision of the subject. Here, we developed a multifunctional contact lens sensor that alleviates some of these limitations since it was developed on an actual ocular contact lens. It was also designed to monitor glucose within tears, as well as intraocular pressure using the resistance and capacitance of the electronic device. Furthermore, in-vivo and in-vitro tests using a live rabbit and bovine eyeball demonstrated its reliable operation. Our developed contact lens sensor can measure the glucose level in tear fluid and intraocular pressure simultaneously but yet independently based on different electrical responses.
Sigma-Aldrich
1-Pyrenebutyric acid, 97%
Pricing and availability is not currently available.
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Supelco
Urea, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.