MilliporeSigma
  • Recurrent rhinovirus infections in a child with inherited MDA5 deficiency.
Recurrent rhinovirus infections in a child with inherited MDA5 deficiency.
The Journal of experimental medicine (2017-06-14)
Ian T Lamborn, Huie Jing, Yu Zhang, Scott B Drutman, Jordan K Abbott, Shirin Munir, Sangeeta Bade, Heardley M Murdock, Celia P Santos, Linda G Brock, Evan Masutani, Emmanuel Y Fordjour, Joshua J McElwee, Jason D Hughes, Dave P Nichols, Aziz Belkadi, Andrew J Oler, Corinne S Happel, Helen F Matthews, Laurent Abel, Peter L Collins, Kanta Subbarao, Erwin W Gelfand, Michael J Ciancanelli, Jean-Laurent Casanova, Helen C Su
ABSTRACT
MDA5 is a cytosolic sensor of double-stranded RNA (ds)RNA including viral byproducts and intermediates. We studied a child with life-threatening, recurrent respiratory tract infections, caused by viruses including human rhinovirus (HRV), influenza virus, and respiratory syncytial virus (RSV). We identified in her a homozygous missense mutation in
Sigma-Aldrich
MISSION® esiRNA, targeting human IFIH1