- Versican Promotes Tumor Progression, Metastasis and Predicts Poor Prognosis in Renal Carcinoma.
Versican Promotes Tumor Progression, Metastasis and Predicts Poor Prognosis in Renal Carcinoma.
Molecular cancer research : MCR (2017-03-01)
Yozo Mitsui, Hiroaki Shiina, Taku Kato, Shigekatsu Maekawa, Yutaka Hashimoto, Marisa Shiina, Mitsuho Imai-Sumida, Priyanka Kulkarni, Pritha Dasgupta, Ryan Kenji Wong, Miho Hiraki, Naoko Arichi, Shinichiro Fukuhara, Soichiro Yamamura, Shahana Majid, Sharanjot Saini, Guoren Deng, Rajvir Dahiya, Koichi Nakajima, Yuichiro Tanaka
PMID28242813
ABSTRACT
The proteoglycan versican (VCAN) promotes tumor progression and enhances metastasis in several cancers; however, its role in clear cell renal cell carcinoma (ccRCC) remains unknown. Recent evidence suggests that VCAN is an important target of chromosomal