MilliporeSigma
  • In vitro and in vivo anti-tumor effects of selected platinum(IV) and dinuclear platinum(II) complexes against lung cancer cells.
In vitro and in vivo anti-tumor effects of selected platinum(IV) and dinuclear platinum(II) complexes against lung cancer cells.
Journal of biological inorganic chemistry : JBIC : a publication of the Society of Biological Inorganic Chemistry (2017-04-20)
Milos Arsenijevic, Marija Milovanovic, Snezana Jovanovic, Natalija Arsenijevic, Bojana Simovic Markovic, Marina Gazdic, Vladislav Volarevic
ABSTRACT
In the present study, cytotoxic effects of cisplatin, the most usually used chemotherapeutic agent, were compared with new designed platinum(IV) ([PtCl
Sigma-Aldrich
1,2-Bis(4-pyridyl)ethane, 99%