MilliporeSigma
  • Effect of interlamellar interactions on shear induced multilamellar vesicle formation.
Effect of interlamellar interactions on shear induced multilamellar vesicle formation.
The Journal of chemical physics (2017-07-25)
Y Kawabata, R Bradbury, S Kugizaki, K Weigandt, Y B Melnichenko, K Sadakane, N L Yamada, H Endo, M Nagao, H Seto
ABSTRACT
Shear-induced multilamellar vesicle (MLV) formation has been studied by coupling the small-angle neutron scattering (SANS) technique with neutron spin echo (NSE) spectroscopy. A 10% mass fraction of the nonionic surfactant pentaethylene glycol dodecyl ether (C
Sigma-Aldrich
Pentaethylene glycol monododecyl ether, BioXtra, ≥98.0% (GC)