MilliporeSigma
  • Isolation, Characterization, and Selection of Molds Associated to Fermented Black Table Olives.
Isolation, Characterization, and Selection of Molds Associated to Fermented Black Table Olives.
Frontiers in microbiology (2017-08-05)
Simona L Bavaro, Antonia Susca, Jens C Frisvad, Maria Tufariello, Agathi Chytiri, Giancarlo Perrone, Giovanni Mita, Antonio F Logrieco, Gianluca Bleve
ABSTRACT
Table olives are one of the most important fermented food in the Mediterranean countries. Apart from lactic acid bacteria and yeasts that mainly conduct the olive fermentation, molds can develop on the brine surface, and can have either deleterious or useful effects on this process. From the food safety point of view, occurring molds could also produce mycotoxins, so, it is important to monitor and control them. In this respect, identification of molds associated to two Italian and two Greek fermented black table olives cultivars, was carried out. Sixty strains were isolated and molecularly identified as
Product Description
Millipore
Chlortetracycline Selective Supplement, suitable for microbiology