MilliporeSigma
  • Assessment of rosehips based on the content of their biologically active compounds.
Assessment of rosehips based on the content of their biologically active compounds.
Journal of food and drug analysis (2017-09-16)
Apurva Bhave, Vera Schulzova, Hana Chmelarova, Libor Mrnka, Jana Hajslova
ABSTRACT
In this study, an in-depth analysis of the unique set of rosehip samples from 71 Rosa genotypes was conducted with the aim to identify the most suitable ones for applications in the food and pharmaceutical industries based on the content of biologically active compounds. In the first part of our experiments, the antioxidant activity was determined by 2,2-diphenyl-1-picrylhydrazyl assay and the genotypes with the highest values were selected for the follow-up analysis. In the second part of experiments, the major classes of biologically active compounds in rosehips such as carotenoids, tocopherols, flavonoids, and triterpenoic acids were further quantified using liquid chromatography-based techniques. Large variation was observed among all the analyzed compounds with intraspecific variation often hiding interspecific or intersectional differences. The compounds studied herein thus do not provide a sharp tool for chemotaxonomic resolution of the genus Rosa. High intraspecific variation indicates the necessity to screen and utilize individual rose genotypes rather than representatives of the species when searching for sources of biologically active compounds. In the final stage of the study, 10 genotypes were selected for further cultivation and use, based on the highest concentrations of the analyzed biologically active compounds.
Sigma-Aldrich
Oleanolic acid, ≥97%
Pricing and availability is not currently available.