MilliporeSigma
  • Combined single-cell quantitation of host and SIV genes and proteins ex vivo reveals host-pathogen interactions in individual cells.
Combined single-cell quantitation of host and SIV genes and proteins ex vivo reveals host-pathogen interactions in individual cells.
PLoS pathogens (2017-06-28)
Diane L Bolton, Kathleen McGinnis, Greg Finak, Pratip Chattopadhyay, Raphael Gottardo, Mario Roederer
ABSTRACT
CD4 T cells harboring HIV-1/SIV represent a formidable hurdle to eradicating infection, and yet their detailed phenotype remains unknown. Here we integrate two single-cell technologies, flow cytometry and highly multiplexed quantitative RT-PCR, to characterize SIV-infected CD4 T cells directly ex vivo. Within individual cells, we correlate the cellular phenotype, in terms of host protein and RNA expression, with stages of the viral life cycle defined by combinatorial expression of viral RNAs. Spliced RNA+ infected cells display multiple memory and activation phenotypes, indicating virus production by diverse CD4 T cell subsets. In most (but not all) cells, progressive infection accompanies post-transcriptional downregulation of CD4 protein, while surface MHC class I is largely retained. Interferon-stimulated genes were also commonly upregulated. Thus, we demonstrate that combined quantitation of transcriptional and post-transcriptional regulation at the single-cell level informs in vivo mechanisms of viral replication and immune evasion.
Sigma-Aldrich
Lectin from Phaseolus vulgaris (red kidney bean), Phytohemagglutinin PHA-P, BioXtra, purified by affinity chromatography, lyophilized powder