MilliporeSigma
CuInS
ACS applied materials & interfaces (2017-06-29)
Yongbo Yang, Li Lin, Lijia Jing, Xiuli Yue, Zhifei Dai
ABSTRACT
A bimodal contrast nanoagent was developed by chelating gadolinium ions to 2-[bis[2-[carboxymethyl-[2-oxo-2-(2-sulfanylethyl-amino)ethyl]amino]ethyl]amino]acetic acid (DTDTPA)-modified CuInS
Sigma-Aldrich
Diethylenetriaminepentaacetic dianhydride, 98%