MilliporeSigma
  • Matrix metalloproteinase processing of PTHrP yields a selective regulator of osteogenesis, PTHrP
Matrix metalloproteinase processing of PTHrP yields a selective regulator of osteogenesis, PTHrP
Oncogene (2017-04-04)
J S Frieling, G Shay, V Izumi, S T Aherne, R G Saul, M Budzevich, J Koomen, C C Lynch
ABSTRACT
Parathyroid hormone-related protein (PTHrP) is a critical regulator of bone resorption and augments osteolysis in skeletal malignancies. Here we report that the mature PTHrP