MilliporeSigma
  • Visible spectrum extended-focus optical coherence microscopy for label-free sub-cellular tomography.
Visible spectrum extended-focus optical coherence microscopy for label-free sub-cellular tomography.
Biomedical optics express (2017-07-19)
Paul J Marchand, Arno Bouwens, Daniel Szlag, David Nguyen, Adrien Descloux, Miguel Sison, Séverine Coquoz, Jérôme Extermann, Theo Lasser
ABSTRACT
We present a novel extended-focus optical coherence microscope (OCM) attaining 0.7 μm axial and 0.4 μm lateral resolution maintained over a depth of 40 μm, while preserving the advantages of Fourier domain OCM. Our system uses an ultra-broad spectrum from a supercontinuum laser source. As the spectrum spans from near-infrared to visible wavelengths (240 nm in bandwidth), we call the system visOCM. The combination of such a broad spectrum with a high-NA objective creates an almost isotropic 3D submicron resolution. We analyze the imaging performance of visOCM on microbead samples and demonstrate its image quality on cell cultures and ex-vivo brain tissue of both healthy and alzheimeric mice. In addition to neuronal cell bodies, fibers and plaques, visOCM imaging of brain tissue reveals fine vascular structures and sub-cellular features through its high spatial resolution. Sub-cellular structures were also observed in live cells and were further revealed through a protocol traditionally used for OCT angiography.
Sigma-Aldrich
Formalin solution, neutral buffered, 10%, histological tissue fixative