MilliporeSigma
  • Microbiota source impact in vitro metabolite colonic production and anti-proliferative effect of spent coffee grounds on human colon cancer cells (HT-29).
Microbiota source impact in vitro metabolite colonic production and anti-proliferative effect of spent coffee grounds on human colon cancer cells (HT-29).
Food research international (Ottawa, Ont.) (2017-06-05)
Angélica María Hernández-Arriaga, B Dave Oomah, Rocio Campos-Vega
ABSTRACT
Human gut flora-mediated non-digestible fraction of spent coffee grounds (hgf-NDSCG) was evaluated for its chemopreventive effect and molecular mechanisms involved on human colon adenocarcinoma HT-29 cell survival using two different microbiota source [lean (L) and overweight (OW)]. The source of human gut flora (hgf) (L or OW) affected the pH of hgf-NDSCG only minimally, but linearly reduced those of hgf-inulin. The variability between lean and overweight microbiota was characterized by the metabolism and/or bioaccessibility of different phenolic metabolites, their intermediate and end products as well as by variable time courses. Apoptosis of colon cancer HT-29 cells depended on the microbiota source with the lean microbiota expressing a low lethal concentration 50 (LC