MilliporeSigma
  • Thiocyanates as attractive redox-active electrolytes for high-energy and environmentally-friendly electrochemical capacitors.
Thiocyanates as attractive redox-active electrolytes for high-energy and environmentally-friendly electrochemical capacitors.
Physical chemistry chemical physics : PCCP (2017-03-07)
Barbara Gorska, Paulina Bujewska, Krzysztof Fic
ABSTRACT
This manuscript reports on the novel insight into the development of high voltage carbon/carbon electrochemical capacitors operating in aqueous solutions of alkali metals and ammonium thiocyanates (KSCN, NaSCN, LiSCN, and NH
Sigma-Aldrich
Lithium thiocyanate hydrate