MilliporeSigma
  • Organotitania-Based Nanostructures as a Suitable Platform for the Implementation of Binary, Ternary, and Fuzzy Logic Systems.
Organotitania-Based Nanostructures as a Suitable Platform for the Implementation of Binary, Ternary, and Fuzzy Logic Systems.
Chemphyschem : a European journal of chemical physics and physical chemistry (2017-04-28)
Andrzej Blachecki, Justyna Mech-Piskorz, Marta Gajewska, Krzysztof Mech, Kacper Pilarczyk, Konrad Szaciłowski
ABSTRACT
Titanium dioxide nanocomposites were synthesized in hierarchical architectures through the use of a 1,4-dihydroxyanthraquinone photosensitizer. In the first step, the dye was either incorporated into the TiO