MilliporeSigma
Dithiocarbamate to modify magnetic graphene oxide nanocomposite (Fe
International journal of biological macromolecules (2017-04-02)
Mohammad Heidarizadeh, Esmail Doustkhah, Sadegh Rostamnia, Parisa Fathi Rezaei, Farshad Darvishi Harzevili, Behzad Zeynizadeh
ABSTRACT
Immobilization of lipase was successfully achieved on the surface of magnetically separable Fe
Sigma-Aldrich
4-Nitrophenyl dodecanoate, ≥98.0% (GC)