MilliporeSigma
Reduction of borazines mediated by low-valent chromium species.
Angewandte Chemie (International ed. in English) (2012-11-20)
Tyler J Carter, Jeff W Kampf, Nathaniel K Szymczak
ABSTRACT
Recharging spent BN fuel: {Cr(CO)(3)} mediates the reduction of borazines by hydride and methyl nucleophiles to generate anionic complexes of dearomatized hexamethylborazine. Subsequent quenching leads to the release of a substituted cyclotriborazane, successfully demonstrating the stepwise reduction of a B=N bond.
Sigma-Aldrich
Hexamethylborazine, ≥95%