MilliporeSigma
  • Asymmetric Catalytic Synthesis of Thiochromenes via an Acyl Transfer-Initiated Cascade.
Asymmetric Catalytic Synthesis of Thiochromenes via an Acyl Transfer-Initiated Cascade.
Organic letters (2016-07-08)
Nicholas A Ahlemeyer, Vladimir B Birman
ABSTRACT
A novel, reagent-free catalytic transformation of α,β-unsaturated thioesters into 2-substituted thiochromenes has been developed, with carbon dioxide as the only byproduct. Amidine-based catalysts, particularly homobenzotetramisole and its analogues, achieve high enantioselectivities and yields in this process.
Sigma-Aldrich
(2R)-2-Phenyl-3,4-dihydro-2H-pyrimido[2,1-b][1,3]benzothiazole, ≥95%