MilliporeSigma
  • Confirmation of β-agonist residues in bovine retina and liver using HPLC-MS/MS and evaluation of matrix-dependent problems.
Confirmation of β-agonist residues in bovine retina and liver using HPLC-MS/MS and evaluation of matrix-dependent problems.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment (2016-12-14)
Patricia Regal, Mónica Díaz-Bao, Rocío Barreiro, Beatriz I Vázquez, Alberto Cepeda
ABSTRACT
β-Agonists or β-adrenergic agonists are synthetic drugs sometimes used for therapeutic purposes in veterinary medicine. However, these compounds also belong to a class of illegal growth promoters that decrease the fat content in farm animals. The purpose of this study was to develop an analytical LC-MS/MS confirmatory method for the detection of nine β-agonists in bovine retina and liver tissues. The obtained CCα values ranged from 0.29 to 1.54 µg kg
Product Description
Supelco
Clenproperol, VETRANAL®, analytical standard
Pricing and availability is not currently available.
Supelco
Cimaterol, VETRANAL®, analytical standard
Pricing and availability is not currently available.