MilliporeSigma
  • Validation of a One-Step Method for Extracting Fatty Acids from Salmon, Chicken and Beef Samples.
Validation of a One-Step Method for Extracting Fatty Acids from Salmon, Chicken and Beef Samples.
Journal of food science (2017-08-24)
Zhichao Zhang, Christine E Richardson, Marie Hennebelle, Ameer Y Taha
ABSTRACT
Fatty acid extraction methods are time-consuming and expensive because they involve multiple steps and copious amounts of extraction solvents. In an effort to streamline the fatty acid extraction process, this study compared the standard Folch lipid extraction method to a one-step method involving a column that selectively elutes the lipid phase. The methods were tested on raw beef, salmon, and chicken. Compared to the standard Folch method, the one-step extraction process generally yielded statistically insignificant differences in chicken and salmon fatty acid concentrations, percent composition and weight percent. Initial testing showed that beef stearic, oleic and total fatty acid concentrations were significantly lower by 9-11% with the one-step method as compared to the Folch method, but retesting on a different batch of samples showed a significant 4-8% increase in several omega-3 and omega-6 fatty acid concentrations with the one-step method relative to the Folch. Overall, the findings reflect the utility of a one-step extraction method for routine and rapid monitoring of fatty acids in chicken and salmon. Inconsistencies in beef concentrations, although minor (within 11%), may be due to matrix effects. A one-step fatty acid extraction method has broad applications for rapidly and routinely monitoring fatty acids in the food supply and formulating controlled dietary interventions.
Sigma-Aldrich
Fatty Acid Extraction Kit, Low Standard, sufficient for 40 extractions
Pricing and availability is not currently available.