MilliporeSigma
  • Adiponitrile-Lithium Bis(trimethylsulfonyl)imide Solutions as Alkyl Carbonate-free Electrolytes for Li
Adiponitrile-Lithium Bis(trimethylsulfonyl)imide Solutions as Alkyl Carbonate-free Electrolytes for Li
Chemphyschem : a European journal of chemical physics and physical chemistry (2017-02-24)
Douaa Farhat, Fouad Ghamouss, Julia Maibach, Kristina Edström, Daniel Lemordant
ABSTRACT
Recently, dinitriles (NC(CH