MilliporeSigma
  • Identification and Characterization of Protein Arginine Methyltransferase 1 in
Identification and Characterization of Protein Arginine Methyltransferase 1 in
The Korean journal of parasitology (2017-05-17)
Eun-Kyung Moon, Hyun-Hee Kong, Yeonchul Hong, Hae-Ahm Lee, Fu-Shi Quan
ABSTRACT
Protein arginine methyltransferase (PRMT) is an important epigenetic regulator in eukaryotic cells. During encystation, an essential process for
Sigma-Aldrich
MISSION® esiRNA, targeting human PRMT1