MilliporeSigma
  • Shape-Controlled Metal-Free Catalysts: Facet-Sensitive Catalytic Activity Induced by the Arrangement Pattern of Noncovalent Supramolecular Chains.
Shape-Controlled Metal-Free Catalysts: Facet-Sensitive Catalytic Activity Induced by the Arrangement Pattern of Noncovalent Supramolecular Chains.
ACS nano (2017-04-18)
Guangwei Geng, Penglei Chen, Bo Guan, Lang Jiang, Zhongfei Xu, Dawei Di, Zeyi Tu, Weichang Hao, Yuanping Yi, Chuncheng Chen, Minghua Liu, Wenping Hu
ABSTRACT
Metal-free catalytic materials have recently received broad attention as promising alternatives to metal-involved catalysts. This is owing to their inherent capability to overcome the inevitable limitations of metal-involved catalysts, such as high sensitivity to poisoning, the limited reserves, high cost and scarcity of metals (especially noble metals), etc. However, the lack of shape-controlled metal-free catalysts with well-defined facets is a formidable bottleneck limiting our understandings on the underlying structure-activity relationship at atomic/molecular level, which thereby restrains their rational design. Here, we report that catalytically active crystals of a porphyrin, 5,10,15,20-tetrakis(pentafluorophenyl)porphyrin, could be shaped into well-defined cubes and sheet-like tetradecahedrons (TDHD), which are exclusively and predominantly enclosed by {101} and {001} facets, respectively. Fascinatingly, compared to the cubes, the TDHDs display substantially enhanced catalytic activity toward water decontamination under visible-light irradiation, although both the architectures have identical crystalline structure. We disclose that such interesting shape-sensitive catalytic activity is ascribed to the distinct spatial separation efficiency of photogenerated electrons and holes induced by single-channel and multichannel charge transport pathways along noncovalent supramolecular chains, which are arranged as parallel-aligned and 2D network patterns, respectively. Our findings provide an ideal scientific platform to guide the rational design of next-generation metal-free catalysts of desired catalytic performances.
Sigma-Aldrich
5,10,15,20-Tetrakis(pentafluorophenyl)porphyrin, ≥90.0% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
2,2,6,6-Tetramethylpiperidine, ≥99%
Pricing and availability is not currently available.