MilliporeSigma
Synthesis and characterization of germanosilicate molecular sieves: GeO
Dalton transactions (Cambridge, England : 2003) (2017-01-31)
Kun Jiao, Zixin Zhang, Xiangyu Xu, Zhi Lv, Jiaqing Song, Cong Lin, Junliang Sun, Mingyuan He, Hermann Gies
ABSTRACT
Many synthesis parameters can influence zeolite crystallization, which include the molar ratio of reagents, water content, temperature, the selection of extraframework cations (organic or inorganic template) and so on. In this paper, two new materials, BUCT-1 with cuboid morphology and BUCT-2 with plate-like morphology were obtained based on the synthesis conditions of ITQ-17 by adjusting the GeO