MilliporeSigma
  • Identification of preoptic sleep neurons using retrograde labelling and gene profiling.
Identification of preoptic sleep neurons using retrograde labelling and gene profiling.
Nature (2017-05-18)
Shinjae Chung, Franz Weber, Peng Zhong, Chan Lek Tan, Thuc Nghi Nguyen, Kevin T Beier, Nikolai Hörmann, Wei-Cheng Chang, Zhe Zhang, Johnny Phong Do, Shenqin Yao, Michael J Krashes, Bosiljka Tasic, Ali Cetin, Hongkui Zeng, Zachary A Knight, Liqun Luo, Yang Dan
ABSTRACT
In humans and other mammalian species, lesions in the preoptic area of the hypothalamus cause profound sleep impairment, indicating a crucial role of the preoptic area in sleep generation. However, the underlying circuit mechanism remains poorly understood. Electrophysiological recordings and c-Fos immunohistochemistry have shown the existence of sleep-active neurons in the preoptic area, especially in the ventrolateral preoptic area and median preoptic nucleus. Pharmacogenetic activation of c-Fos-labelled sleep-active neurons has been shown to induce sleep. However, the sleep-active neurons are spatially intermingled with wake-active neurons, making it difficult to target the sleep neurons specifically for circuit analysis. Here we identify a population of preoptic area sleep neurons on the basis of their projection target and discover their molecular markers. Using a lentivirus expressing channelrhodopsin-2 or a light-activated chloride channel for retrograde labelling, bidirectional optogenetic manipulation, and optrode recording, we show that the preoptic area GABAergic neurons projecting to the tuberomammillary nucleus are both sleep active and sleep promoting. Furthermore, translating ribosome affinity purification and single-cell RNA sequencing identify candidate markers for these neurons, and optogenetic and pharmacogenetic manipulations demonstrate that several peptide markers (cholecystokinin, corticotropin-releasing hormone, and tachykinin 1) label sleep-promoting neurons. Together, these findings provide easy genetic access to sleep-promoting preoptic area neurons and a valuable entry point for dissecting the sleep control circuit.
Sigma-Aldrich
SIGMAFAST Fast Red TR/Naphthol AS-MX Tablets, tablet, To prepare 10 mL
Pricing and availability is not currently available.