MilliporeSigma
  • Inhibition of the endogenous volume-regulated anion channel (VRAC) in HEK293 cells by acidic di-aryl-ureas.
Inhibition of the endogenous volume-regulated anion channel (VRAC) in HEK293 cells by acidic di-aryl-ureas.
The Journal of membrane biology (2004-01-16)
N Hélix, D Strøbaek, B H Dahl, P Christophersen
ABSTRACT
The endogenous volume-regulated anion channel (VRAC) from HEK293 cells was pharmacologically characterized using the whole-cell patch-clamp technique. Under isotonic conditions a small (1.3 nS), Ca(2+)-independent Cl conductance was measured. However, swelling at 75% tonicity activated a VRAC identified as an outward-rectifying anion current ( P(l) > P(Cl) > P(gluconate)), which was ATP-dependent and showed inactivation at positive potentials. Activation of this current followed a sigmoid time course, reaching a plateau conductance of 42.6 nS after 12-15 min ( t(1/2) = 7 min). The pharmacology of this VRAC was investigated using standard Cl(-)-channel blockers (NPPB, DIDS, and tamoxifen) as well as a new group (acidic di-aryl ureas) of Cl(-)-channel blockers (NS1652, NS3623, NS3749, and NS3728). The acidic di-aryl ureas were originally synthezised for inhibition of the human erythrocyte Cl(-) conductance in vivo. NS3728 was the most potent VRAC blocker in this series ( IC(50) = 0.40 micro M) and even more potent than tamoxifen (2.2 micro M). NS3728 accelerated channel inactivation at positive potentials. These results show that acidic di-aryl ureas constitute a promising starting point for the synthesis of potent inhibitors of VRAC.
Sigma-Aldrich
NS3728, ≥98% (HPLC)
Pricing and availability is not currently available.