MilliporeSigma
  • Up-regulation of long non-coding RNA XLOC_010235 regulates epithelial-to-mesenchymal transition to promote metastasis by associating with Snail1 in gastric cancer.
Up-regulation of long non-coding RNA XLOC_010235 regulates epithelial-to-mesenchymal transition to promote metastasis by associating with Snail1 in gastric cancer.
Scientific reports (2017-05-28)
Yu-Yi Liu, Ze-Hong Chen, Jian-Jun Peng, Jia-Lin Wu, Yu-Jie Yuan, Er-Tao Zhai, Shi-Rong Cai, Yu-Long He, Wu Song
ABSTRACT
We previously performed long non-coding RNA (lncRNA) expression microarray analyses to identify novel indicators for gastric cancer (GC) metastasis and prognosis in which we identified lncRNA XLOC_010235 (XLOC) as a candidate RNA. However, XLOC_010235 molecular mechanism of action remains unclear. Gain and loss of function approaches were used to investigate the biological role of XLOC in vitro. The effects of XLOC on cell viability were assessed by CCK-8 proliferation assays. Real-time PCR, western-blot and immunofluorescence were used to evaluate the mRNA and protein expression of Snail and multiple EMT related molecules. The positive XLOC/Snail1 interaction was identified and verified by immunohistochemistry assay and bivariate correlation analysis. Ectopic expression of XLOC facilitate cell viability, migration and invasion, leading to the acceleration of metastasis, while depletion of XLOC expression hindered cell migration and invasion. Moreover, over-expression of XLOC was found to play a important role in epithelial-to-mesenchymal transition (EMT) through the regulation of E-cadherin, N-cadherin and Vimentin expression, in which transcriptional factor Snail1 was involved. These results advance our understanding of the role of lncRNA XLOC_010235 as a active regulator of EMT by associating with Snail1, which may help in the development of new therapeutics.
Sigma-Aldrich
MISSION® esiRNA, targeting human SNAI1