MilliporeSigma
  • Chemical Structure of the Lipid A component of Pseudomonas sp. strain PAMC 28618 from Thawing Permafrost in Relation to Pathogenicity.
Chemical Structure of the Lipid A component of Pseudomonas sp. strain PAMC 28618 from Thawing Permafrost in Relation to Pathogenicity.
Scientific reports (2017-05-21)
Han-Gyu Park, Ganesan Sathiyanarayanan, Cheol-Hwan Hwang, Da-Hee Ann, Jung-Ho Kim, Geul Bang, Kyoung-Soon Jang, Hee Wook Ryu, Yoo Kyung Lee, Yung-Hun Yang, Yun-Gon Kim
ABSTRACT
Climate change causes permafrost thawing, and we are confronted with the unpredictable risk of newly discovered permafrost microbes that have disease-causing capabilities. Here, we first characterized the detailed chemical structure of the lipid A moiety from a Pseudomonas species that was isolated from thawing arctic permafrost using MALDI-based mass spectrometric approaches (i.e., MALDI-TOF MS and MALDI-QIT-TOF MS
Product Description
Millipore
Pseudomonas agar base, NutriSelect® Plus, according to ISO 11059:2009, according to ISO 16266:2006, according to ISO 13720:2010, powder, selective for Pseudomonas spp., pack of 500 g, suitable for microbiology
Millipore
Pseudomonas Isolation Agar, NutriSelect® Plus, powder, selective for Pseudomonas spp., pack of 500 g, suitable for microbiology