MilliporeSigma
  • Distinct ON/OFF fluorescence signals from dual-responsive activatable nanoprobes allows detection of inflammation with improved contrast.
Distinct ON/OFF fluorescence signals from dual-responsive activatable nanoprobes allows detection of inflammation with improved contrast.
Biomaterials (2017-04-24)
Mathieu L Viger, Guillaume Collet, Jacques Lux, Viet Anh Nguyen Huu, Monica Guma, Alexandra Foucault-Collet, Jason Olejniczak, Shivanjali Joshi-Barr, Gary S Firestein, Adah Almutairi
ABSTRACT
Visualization of biochemical changes associated with disease is of great clinical significance, as it should allow earlier, more accurate diagnosis than structural imaging, facilitating timely clinical intervention. Herein, we report combining stimuli-responsive polymers and near-infrared fluorescent dyes (emission max: 790 nm) to create robust activatable fluorescent nanoprobes capable of simultaneously detecting acidosis and oxidative stress associated with inflammatory microenvironments. The spectrally-resolved mechanism of fluorescence activation allows removal of unwanted background signal (up to 20-fold reduction) and isolation of a pure activated signal, which enables sensitive and unambiguous localization of inflamed areas; target-to-background ratios reach 22 as early as 3 h post-injection. This new detection platform could have significant clinical impact in early detection of pathologies, individual tailoring of drug therapy, and image-guided tumor resection.
Sigma-Aldrich
2-Methoxypropene, 97%
Pricing and availability is not currently available.