MilliporeSigma
  • High pressure processing for dark-firm-dry beef: effect on physical properties and oxidative deterioration during refrigerated storage.
High pressure processing for dark-firm-dry beef: effect on physical properties and oxidative deterioration during refrigerated storage.
Asian-Australasian journal of animal sciences (2016-07-08)
Dicky Tri Utama, Seung Gyu Lee, Ki Ho Baek, Woon Si Chung, In Ae Chung, Jung Tae Jeon, Sung Ki Lee
ABSTRACT
Study on the application of high pressure processing (HPP) for dark-firm-dry (DFD) beef was conducted to observe whether HPP has any impact on physical properties and to evaluate oxidative deterioration during refrigerated storage under vacuum. The HPP increased meat pH by 0.1 to 0.2 units and the tenderness of cooked DFD beef significantly with no significant effects on meat texture profile. The stability of meat pH was well maintained during refrigerated storage under vacuum. No clear effects were found on the activity of catalase and superoxide dismutase, however, glutathione peroxidase activity was significantly reduced by high pressure. HPP and storage time resulted in aroma changes and the increasing amount of malondialdehyde and metmyoglobin relative composition. Although the increasing amount of malondialdehyde content, metmyoglobin formation and aroma changes in HPP-treated samples could not be avoided, HPP at 200 MPa increased L* and a* values with less discoloration and oxidative deterioration during storage.
Product Description
Supelco
Vial crimp seals, BiMetal (magnetic), 20 mm, with PTFE/silicone septa, pkg 100, blue seal, PTFE/silicone septa, diam. × thickness 20 mm × 3 mm
Pricing and availability is not currently available.