MilliporeSigma
Determination of lithium and transition metals in Li
Electrophoresis (2016-11-22)
Britta Vortmann-Westhoven, Constantin Lürenbaum, Martin Winter, Sascha Nowak
ABSTRACT
In this work, we present a novel electrophoretic method that was developed for the determination of lithium and transition metals in LiNi
Product Description
Fly ash (trace elements), BCR®, certified reference material