MilliporeSigma
  • Phase separation in ceramide[NP] containing lipid model membranes: neutron diffraction and solid-state NMR.
Phase separation in ceramide[NP] containing lipid model membranes: neutron diffraction and solid-state NMR.
Soft matter (2017-02-23)
Annett Schroeter, Sören Stahlberg, Barbora Školová, Stefan Sonnenberger, Adina Eichner, Daniel Huster, Kateřina Vávrová, Thomas Hauß, Bodo Dobner, Reinhard H H Neubert, Alexander Vogel
ABSTRACT
The stratum corneum is the outermost layer of the skin and protects the organism against external influences as well as water loss. It consists of corneocytes embedded in a mixture of ceramides, fatty acids, and cholesterol in a molar ratio of roughly 1 : 1 : 1. The unique structural and compositional arrangement of these stratum corneum lipids is responsible for the skin barrier properties. Many studies investigated the organization of these barrier lipids and, in particular, the exact conformation of ceramides. However, so far no consensus has been reached. In this study, we investigate a model system comprised of N-(non-hydroxy-tetracosanoyl)-phytosphingosine/cholesterol/tetracosanoic acid (CER[NP]-C24/CHOL/TA) at a 1 : 1 : 1 molar ratio using neutron diffraction and
Sigma-Aldrich
(2S,3S,4R)-2-Amino-1,3,4-octadecanetriol, AldrichCPR