MilliporeSigma
  • Colorimetric assay for ?-lactamase activity using cocktail of penicillin and 4-(2-pyridylazo)resorcinol (PAR)?2Hg2+ complex
Colorimetric assay for ?-lactamase activity using cocktail of penicillin and 4-(2-pyridylazo)resorcinol (PAR)?2Hg2+ complex
Dyes and Pigments (2017)
Suji Lee